Recombinat Protein,Human NKG2A CD159a Protein, Fc Tag,Human NKG2A, Fc Tag (NKA-H526c) is expressed from human 293 cells (HEK293). It contains AA Pro 94 - Leu 233 (Accession P26715-1).This protein carries a human IgG1 Fc tag at the N-terminus. The pr 1 * 100 µG
Recombinat Protein,Human NKG2A CD159a Protein, Fc Tag,Human NKG2A, Fc Tag (NKA-H526c) is expressed from human 293 cells (HEK293). It contains AA Pro 94 - Leu 233 (Accession P26715-1).This protein carries a human IgG1 Fc tag at the N-terminus. The pr 1 * 100 µG