PC certified clean validation bottles, Nalgene®
5 ml, PC certified clean validation bottles
Catalog # 216-2287
Supplier: Thermo Fisher Scientific
PC certified clean validation bottles, Nalgene®
Catalog # 216-2287
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
The Nalgene™ multi-tiered portfolio provides bioproduction facilities with a variety of high-quality, sterile storage and transport solutions. Choose the appropriate product necessary to suit any application, through the step-down process and into the cleanroom.
- Polycarbonate construction
- Sterile
- Clear finish
- Certified clean
These bottles can reduce the risk of contamination.
Certifications: Validated to USP 788 and <0,5 EU/ml endotoxin.
Specifications
- Capacità:5 ml
- Descrizione:PC certified clean validation bottles
- Seq:01
- Cat. No.:216-2287
- Environmentally Preferable:Produzione a basso impatto
- Certificazione ambientale:NAR Certificate of Retirement