Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 SFr.
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MABSol® Biotinylated Human TNF-alpha, epitope tag free, primary amine labeling (active trimer) (MALS verified) (TNA-H8211) is expres null
Catalog # ACRBTNA-H8211-25UG
Supplier:
ACRO Biosystems
Expand image
MABSol® Biotinylated Human TNF-alpha, epitope tag free, primary amine labeling (active trimer) (MALS verified) (TNA-H8211) is expres null
Catalog # ACRBTNA-H8211-25UG
Supplier:
ACRO Biosystems
Frequently Bought Together
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR®, Pipette sierologiche, Standard Line
Supplier: VWR Collection
Provette per centrifuga, Falcon®
Supplier: Corning
Pipette sierologiche, Stripette™
Supplier: Corning
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
VWR®, Piastra Petri, PS
Supplier: VWR Collection
VWR®, Contenitori per campioni dritti con tappo a vite
Supplier: VWR Collection
VWR®, Provette e strip per PCR, 0,2 ml
Supplier: VWR Collection
VWR®, pipette di trasferimento, alte prestazioni
Supplier: VWR Collection
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
VWR®, Pipette Pasteur, Standard Line
Supplier: VWR Collection
Provette per microcentrifuga Eppendorf® Protein LoBind®
Supplier: EPPENDORF
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF