Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Dumont vessel cannulation forceps

Dumont vessel cannulation forceps

1 mm OD tubing size, Inox, 110 mm, Angled
Catalog # FSCI11282-11
Marketsource
Supplier:   FINE SCIENCE TOOLS
Dumont vessel cannulation forceps
Catalog # FSCI11282-11
Supplier:  FINE SCIENCE TOOLS
Marketsource
Dumont vessel cannulation forceps
Dumont vessel cannulation forceps

1 mm OD tubing size, Inox, 110 mm, Angled

Price
SFr. 167.19
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Descrizione:
    1 mm OD tubing size
  • Lunghezza:
    110 mm
  • Materiale:
    Inox
  • Seq:
    02
  • Tip shape:
    Angled
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.