Standard hazard warning signs
Warning - low-hanging obstacles - risk of impact (W020), In PE, 100×87 mm
Codice catalogo 116-0946
Nome del fornitore: Brady
Vai a
Product Details & Documents
Warning signs must have a black pictogram on a yellow triangle with a black border. Hazard signs must be displayed to warn people to be careful or to take precaution where hazards exist.
- Used for a wide range of applications
- Resistance to most chemical products and solvents
- Resistance against extreme temperatures
- Thickness: 1,5 mm
- Non-adhesive
Specifiche
- Descrizione:Warning - low-hanging obstacles - risk of impact (W020)
- Dimensioni:100×87 mm
- Materiale:In PE
- Seq:23
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



