Objectives for REVOLVE 3M microscope
POL U-Plan fluorite objective, 4×, 17 mm
Catalog # ECHLJMAG0046
Supplier: DISCOVER ECHO
Jump to:
Product Details & Documents
Documents
ECHO Meet Revolve
Product Literature
Specifications
- Ingrandimento:4×
- Descrizione:POL U-Plan fluorite objective
- Seq:05
- Distanza di lavoro:17 mm