Condensers for REVOLVE 3M microscope
Condensatore universale swing out NA 0,9 WD 0,17 mm
Catalog # ECHLJCND2003
Supplier: DISCOVER ECHO
Jump to:
Product Details & Documents
Documents
ECHO Meet Revolve
Product Literature
Specifications
- Descrizione:Condensatore universale swing out NA 0,9 WD 0,17 mm
- Seq:04