Orders LinkContactUsLinkComponent
MultiRack tipstations, Axygen®
MultiRack tipstations, Axygen®
0,1 - 10 µl, -, Stack: 960 tips/stack, 5 stacks/case
Codice catalogo AXYGMR-10XT-TS
Nome del fornitore:   Corning
MultiRack tipstations, Axygen®
Codice catalogo AXYGMR-10XT-TS
Nome del fornitore:  Corning
MultiRack tipstations, Axygen®
MultiRack tipstations, Axygen®
0,1 - 10 µl, -, Stack: 960 tips/stack, 5 stacks/case
Price
SFr. 363.00
Log in per disponibilità
1 * 4.800 ST
Product Details & Documents
The Axygen® MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in bulk, racked, filter and reload packaging styles to meet the variety of needs in the lab setting.
  • Detectable DNase/RNase/Human DNA/PCR inhibition free
  • Non pyrogenic
  • Manufactured in an ISO certified facility in the USA
Axygen® MultiRack tipstation is packaged in a format with 192 tips per layer and five layers total and can be used with 8 or 12 channel pipettors. This packaging style provides a convenient solution for lab applications such as ELISA, PCR, and NGS library preparation, where thousands of tips are consumed. Axygen® MultiRack tipstation is available with 10 and 200 µl, sterile or non sterile.
Informazioni per l'ordine: Axygen® MultiRack pipette tips are also available in bulk, hinged rack, filter tips and reload configurations.
Specifiche
  • Confezione:
    Stack: 960 tips/stack, 5 stacks/case
  • Sterili:
    -
  • Volume:
    0,1 - 10 µl
  • Seq:
    01