FLUIDX 96-FORMAT, 0.7ML INTERNAL THREAD, NEXT-GEN DUAL-CODED TUBE, 2D CODE AND HUMAN READABLE NUMBER ON BASE, CAPPED, 10 RACKS PER C ASE HIGHBASE RACK, WITH TUBELOCK EMPTY RACK PART NUMBER: 66-51016 1 * 960 ST
Catalog # MSPP66-62319-Y6
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together