Polysorbate 80 FCC
Product Details & Documents
Polysorbate 80, FCC is used as a wetting agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Kosher certified
Storage temperature - Keep tightly closed, protect from light.
- Numero CAS:9005-65-6
- Numero MDL:MFCD00082107
- Punto di fusione:100 °C
- Punto di infiammabilità:113 °C (coppa chiusa)
- temperatura di conservazione:Ambiente
- EINECS:500-019-9
Specifiche
Risultati dei test sulle specifiche
- Assay:65% Min.
Specifiche
- Tipo di conf.:Poly drum
- Conf:20 L
- Cat. No.:SPCMP1179-20LT
Specifiche
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



