Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Sleeve samplers, U-D Master

Tip for U-D sleeve sampler, Acciaio inossidabile 316L, 0,3 ml
  SAMP1500W-03
Sleeve samplers, U-D Master
  SAMP1500W-03
Sleeve samplers, U-D Master
Sleeve samplers, U-D Master

Tip for U-D sleeve sampler, Acciaio inossidabile 316L, 0,3 ml

 
SFr. 162.00
   
1 * 1 ST
 

The U-D sleeve sampler was developed for accurate unit dose samples (small volume).

  • Quick and easy accurate unit dose sampling
  • Different volumes can be taken using the same sampler
  • Simply change the tip for a different volume

Designed for use with free flowing powders and granules.

 
  • Volume:
    0,3 ml
  • Descrizione:
    Tip for U-D sleeve sampler
  • Materiale:
    Acciaio inossidabile 316L
  • Seq:
    08