Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Sleeve samplers, U-D Master

Sleeve samplers, U-D Master

Tip for U-D sleeve sampler, Acciaio inossidabile 316L, 0,8 ml
Codice catalogo SAMP1500W-08
Nome del fornitore:   Sampling Systems
Sleeve samplers, U-D Master
Codice catalogo SAMP1500W-08
Nome del fornitore:  Sampling Systems
Sleeve samplers, U-D Master
Sleeve samplers, U-D Master

Tip for U-D sleeve sampler, Acciaio inossidabile 316L, 0,8 ml

Price
SFr. 162.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents

The U-D sleeve sampler was developed for accurate unit dose samples (small volume).

  • Quick and easy accurate unit dose sampling
  • Different volumes can be taken using the same sampler
  • Simply change the tip for a different volume

Designed for use with free flowing powders and granules.

Specifiche
  • Volume:
    0,8 ml
  • Descrizione:
    Tip for U-D sleeve sampler
  • Materiale:
    Acciaio inossidabile 316L
  • Seq:
    10
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.