Handheld digital microscopes, Dino-Lite Wireless
Product Details & Documents
These microscopes from the Dino-Lite Universal series work wired via USB or wireless with the integrated WF-20 Wireless adapter.
- Wireless and wired in one device
- Work with different operating systems: iOS, Android or Windows
- USB 2.0, enhanced 1,3 MP EDGE sensor
- Integrated polariser greatly reduces glare on reflective surfaces
- Extensive measurement functionality
- Flexible LED control (FLC): Additional LED controls within the included software for adjusting LED brightness and on/off toggle by quadrant
Models with Automatic Magnification Reading (AMR): Allows the included software to read the magnification setting from the microscope device, simplifying the measuring process.
Models with Extended Depth of Field (EDOF): Automatically capture and combine multiple images at different focus levels to produce a final image with greater depth of field.
Models with Extended Dynamic Range (EDR): Automatically capture and combine multiple images at different exposure levels to create an image with greater dynamic range.
Ideal for inspections in hard-to-reach places in factories, in the field, in workshops or in greenhouses.
Documents
Dino-Lite Powerful Portable Versatile Dino-Lite Software Dino-Lite Wireless, WF series
Letteratura sul prodotto
Specifiche
- Ingrandimento:20 - 220×
- Modello:WF4915ZT
- Risoluzione:1,3 MP
- Descrizione:Wireless microscope
- Seq:05
- Caratteristiche aggiuntive:AMR / EDOF / EDR
Accessories
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Sturdy high-end stand for Dino-Lite microscopes, RK-10A, incl. HD-P1 holder and RK-10-EX horizontal armSturdy high-end stand for Dino-Lite microscopes, RK-10A, incl. HD-P1 holder and RK-10-EX horizontal arm630-2517Log in per disponibilità1 * 1 ST - 323.00 SFr.
Informazioni sul prodotto
- Codice catalogoL×P×ADisponibilitàPrice
- Tabletop stand for handheld Dino-Lite microscopes, RK-06-A, incl. extension arm RK-10-EXTabletop stand for handheld Dino-Lite microscopes, RK-06-A, incl. extension arm RK-10-EXIDCPRK-06A150×220×270 mmLog in per disponibilità1 * 1 ST - 240.00 SFr.
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Foot pedal, enables users to take a picture with their footFoot pedal, enables users to take a picture with their foot630-1821Log in per disponibilità1 * 1 ST - 60.10 SFr.