0.00 SFr.
Inserisci ordine
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
Orders Link
ContactUsLinkComponent
Obiettivi del microscopio
Objectives for Rebel Microscopes
Objectives for Rebel Microscopes
Plan achromat phase, N.A. 0,4, 20×, 7,9 mm
ECHLRMAG0213
:
DISCOVER ECHO
Objectives for Rebel Microscopes
ECHLRMAG0213
:
DISCOVER ECHO
Objectives for Rebel Microscopes
Plan achromat phase, N.A. 0,4, 20×, 7,9 mm
SFr. 703.00
1 * 1 ST
-
+
Discover Echco Hybrid Microscope
ECHO Rebel Microscope Product Range
ECHO Rebel Microscope Specification
Ingrandimento:
20×
Descrizione:
Plan achromat phase, N.A. 0,4
Seq:
23
Distanza di lavoro:
7,9 mm
Hybrid microscope (upright + inverted), ECHO Rebel
#: 630-3346
: DISCOVER ECHO