Orders LinkContactUsLinkComponent
Inserts for trays H 44 series | Avantor
Inserts for trays H 44 series
ChipGuard™ 2000 antistatic tray inserts, thickness 5 - 26 mm, bianco, Tyvek®, Yes
Katalognr. ENTEH44-001D
Anbieter:   ENTEGRIS
Inserts for trays H 44 series
Katalognr. ENTEH44-001D
Anbieter:  ENTEGRIS
Inserts for trays H 44 series
ChipGuard™ 2000 antistatic tray inserts, thickness 5 - 26 mm, bianco, Tyvek®, Yes
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 SET
Product Details & Documents
Nest between H 44 trays and/or covers.
  • Precut to tray size
  • Inserts provide a softer surface providing protection for more sensitive components
  • Inserts are not intended to limit part migration
  • Inserts with antistatic coatings are not recommended for use when storing devices for extended periods
Technische Daten
  • Colore:
    bianco
  • Descrizione:
    ChipGuard™ 2000 antistatic tray inserts, thickness 5 - 26 mm
  • Materiale:
    Tyvek®
  • Seq:
    01
  • Standard:
    Yes