Inserts for trays H 44 series
ChipGuard™ 2000 antistatic tray inserts, thickness 5 - 26 mm, bianco, Tyvek®, Yes
Katalognr. ENTEH44-001D
Anbieter: ENTEGRIS
Direkt zu
Product Details & Documents
Nest between H 44 trays and/or covers.
- Precut to tray size
- Inserts provide a softer surface providing protection for more sensitive components
- Inserts are not intended to limit part migration
- Inserts with antistatic coatings are not recommended for use when storing devices for extended periods
Technische Daten
- Colore:bianco
- Descrizione:ChipGuard™ 2000 antistatic tray inserts, thickness 5 - 26 mm
- Materiale:Tyvek®
- Seq:01
- Standard:Yes