Orders LinkContactUsLinkComponent
Pierce™ Fish Serum Blocking Buffer,Thermo Scientific
Fish Serum blocking Buffer, contains steelhead salmon serum in phosphate buffered saline and 0,1% sodium azide, 1.500 ml
Codice catalogo PIER37527X3
Nome del fornitore:   Thermo Fisher Scientific
Pierce™ Fish Serum Blocking Buffer,Thermo Scientific
Codice catalogo PIER37527X3
Nome del fornitore:  Thermo Fisher Scientific
Numero CAS:  
Pierce™ Fish Serum Blocking Buffer,Thermo Scientific
Fish Serum blocking Buffer, contains steelhead salmon serum in phosphate buffered saline and 0,1% sodium azide, 1.500 ml
Numero CAS:  
Price
1 060.00 SFr.
Log in per disponibilità
1 * 1.500 mL
Product Details & Documents
This Fish Serum blocking buffer is steelhead salmon serum in PBS that is especially useful as a blocking agent in IHC and other detection methods involving mammalian samples.
  • Non mammalian
  • Easy to use
  • Convenient, filtered and stabilized in PBS
Fish Serum blocking buffer is fish serum, it is effective in stabilising solutions and samples for antibody-binding interactions while at the same time minimizing the possibility of cross-reaction with mammalian sample components. The product is also especially effective in various cellular imaging protocols based on visible and near infrared fluorescence detection. The blocker has been used to decrease background and increase signal-to-noise ratios with near IR fluorescent probes and is validated for use with the LI-COR Odyssey™ Infrared Imaging System.
Specifiche
Specifiche
  • Conf:
    1.500 ml
  • Descrizione:
    Fish Serum blocking Buffer, contains steelhead salmon serum in phosphate buffered saline and 0,1% sodium azide