Caps for sample tubes, tamper evident
Lip seal screw cap, not tamper evident, 16,6 mm, bianco
Catalog # SIMPT552W
Supplier: Simport Scientific
Jump to:
Product Details & Documents
Caps made of PE feature a long skirt and a super fast thread design allowing them to be removed or sealed with a single turn. Caps are designed with flexible sealing lip which provides leakproof seal.
Caution: *I prodotti in questa pagina non rientrano nell'ambito della Direttiva UE 2019/904
Specifications
- Per provette:16,6 mm
- Colore:bianco
- Descrizione:Lip seal screw cap, not tamper evident
- Seq:05