ContactUsLinkComponent
GG-12-1.5-PLL: 12 mm diameter, #1.5 thickness, 90 pieces of CE certified crystal clear German glass coverslips with poly-L-lysine coating on both sides for cell culture and confocal fluorescence microscopy, sealed 15 pieces per bag x 6 = 90, steriliz 1 * 90 ST
Codice catalogo MSPPGG-12-1.5-PLL
Nome del fornitore:  MARKET SOURCE PART PROCESS
GG-12-1.5-PLL: 12 mm diameter, #1.5 thickness, 90 pieces of CE certified crystal clear German glass coverslips with poly-L-lysine coating on both sides for cell culture and confocal fluorescence microscopy, sealed 15 pieces per bag x 6 = 90, steriliz 1 * 90 ST
Codice catalogo MSPPGG-12-1.5-PLL
Nome del fornitore:  MARKET SOURCE PART PROCESS
Frequently Bought Together