Labels for BradyJet J2000 Lab printer
Printer labels, B-7425J, J20-179-7425J, PP, bianco, J2000-Lab, 25×25 mm, 1500 labels/roll
Catalog # BRDY150025
Supplier: Brady
Jump to:
Product Details & Documents
These PP BradyJet J2000 Lab printer labels come in matching sizes and shapes for most sample containers including tubes, vials, cryo containers, dishes, plates and general glassware.
- Designed to stay attached and remain legible
- Resist liquid nitrogen, freezers, hot water baths and autoclaves
They can also resist common laboratory chemicals like acetone, ethanol, toluene, xylene and IPA.
Specifications
- Stampante:J2000-Lab
- Tipo:B-7425J
- Colore:bianco
- Descrizione:Printer labels
- Dimensioni:25×25 mm
- Materiale:PP
- Seq:07
- Modello:J20-179-7425J
- Confezione:1500 labels/roll