Labels for BradyJet J2000 Lab printer
Printer labels, B-7425J, J20-59-7425J, PP, bianco, J2000-Lab, 25×12 mm, 2000 labels/roll
Codice catalogo BRDY150006
Nome del fornitore: Brady
Vai a
Product Details & Documents
These PP BradyJet J2000 Lab printer labels come in matching sizes and shapes for most sample containers including tubes, vials, cryo containers, dishes, plates and general glassware.
- Designed to stay attached and remain legible
- Resist liquid nitrogen, freezers, hot water baths and autoclaves
They can also resist common laboratory chemicals like acetone, ethanol, toluene, xylene and IPA.
Documents
Specifiche
- Stampante:J2000-Lab
- Tipo:B-7425J
- Colore:bianco
- Descrizione:Printer labels
- Dimensioni:25×12 mm
- Materiale:PP
- Seq:04
- Modello:J20-59-7425J
- Confezione:2000 labels/roll