Orders LinkContactUsLinkComponent
VWR® Evaporating Flasks
VWR® Evaporating Flasks
100 ml, 29/32 NS
Codice catalogo 531-1099
Nome del fornitore:   VWR Collection
VWR® Evaporating Flasks
Codice catalogo 531-1099
Nome del fornitore:  VWR Collection
VWR® Evaporating Flasks
VWR® Evaporating Flasks
100 ml, 29/32 NS
Price
22.80 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Borosilicate 3.3 glass.
  • Very good chemical resistance
  • High temperature resistance and high resistance to thermal shock
  • White labelling area and imprinted catalogue number
These pear-shaped rotary evaporating flasks are produced from thick walled borosilicate glass tubing. With standard ground socket, suitable for all standard rotary evaporators.
Specifiche
  • Cono femmina:
    29/32 NS
  • Capacità:
    100 ml
  • Seq:
    01