VWR®, Testine Splash, Stutzer, Dritte
29/32 NS, 14/23 NS
Supplier: VWR Collection
Jump to:
Product Details & Documents
Borosilicate 3.3 glass.
- Very good chemical resistance
- High temperature resistance and high resistance to thermal shock
- Straight, with standard ground joint and socket
These splash heads, according to Stutzer, can be placed directly onto the distillation flask to intercept droplets.
Specifications
- Dimensione cono:14/23 NS
- Cono femmina:29/32 NS
- Seq:02