Orders LinkContactUsLinkComponent
VWR®, Testine Splash, Stutzer, Dritte
VWR®, Testine Splash, Stutzer, Dritte
14/23 NS, 14/23 NS
Codice catalogo 201-1398
Nome del fornitore:   VWR Collection
VWR®, Testine Splash, Stutzer, Dritte
Codice catalogo 201-1398
Nome del fornitore:  VWR Collection
VWR®, Testine Splash, Stutzer, Dritte
VWR®, Testine Splash, Stutzer, Dritte
14/23 NS, 14/23 NS
Price
SFr. 47.80
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Borosilicate 3.3 glass.
  • Very good chemical resistance
  • High temperature resistance and high resistance to thermal shock
  • Straight, with standard ground joint and socket
These splash heads, according to Stutzer, can be placed directly onto the distillation flask to intercept droplets.
Specifiche
  • Dimensione cono:
    14/23 NS
  • Cono femmina:
    14/23 NS
  • Seq:
    01