Zero pressure weather caps
Zero pressure weather caps, 1422 mm height, 305 mm nominal diameter,15,9 kg shipping weight
Catalog # LBCP5622100
Supplier: LABCONCO
Jump to:
Product Details & Documents
Weathercaps are constructed of strong, corrosion-resistant PVC.
Caps fits atop standard PVC duct, and permit vertical discharge of effluent air above roofline for dispersion away from the building. Caps add little static pressure to the exhaust system.
Specifications
- Descrizione:Zero pressure weather caps, 1422 mm height, 305 mm nominal diameter,15,9 kg shipping weight
- Seq:02