Absorbent socs, Allwik Universal
43 L, 6 pieces/box, 76×244 mm
Codice catalogo BRDY813790
Nome del fornitore: Brady
Vai a
Product Details & Documents
These Allwik-Socs are used for absorbing leaks and spills around machines.
- Absorbs a variety of industrial liquids including aggressive chemicals
- Flexible design allows the soc to fit around any machine, corner or curve
- Filled with universal meltblown polypropylene
Specifiche
- Capacità:43 L
- Confezione:6 pieces/box
- Ø×L:76×244 mm



