[EN]BACTOBEADS E. COLI GFP HOST 1 * 5 pezzi
Catalog # EDVO728
Supplier: EDVOTEK
Frequently Bought Together
2-Propanolo ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acqua, LiChrosolv® per cromatografia, per LC-MS, Supelco®
Supplier: Merck
Metanolo ≥99.8%, HiPerSolv CHROMANORM®, grado isocratico per HPLC
Supplier: VWR Chemicals
n-Esano ≥97%, HiPerSolv CHROMANORM® per HPLC
Supplier: VWR Chemicals
Acqua, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Supplier: VWR Chemicals
Acqua, HiPerSolv CHROMANORM® per HPLC
Supplier: VWR Chemicals
2-Propanolo ≥99.8%, HiPerSolv CHROMANORM® per HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, grado gradiente per HPLC
Supplier: VWR Chemicals
Etanolo, GPR RECTAPUR®
Supplier: VWR Chemicals
Acqua, HiPerSolv CHROMANORM®, super gradient grade per HPLC, adatto per UPLC/UHPLC
Supplier: VWR Chemicals
Etanolo 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
VWR®, Pre-weighed dehydrated culture media in sachets
Supplier: VWR Chemicals