Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Magnetic plates

Magnetic plates

Super magnetic plate, 8×12 cm
Codice catalogo OZBIMF10000
Nome del fornitore:   OZ BIOSCIENCES
Magnetic plates
Codice catalogo OZBIMF10000
Nome del fornitore:  OZ BIOSCIENCES
Magnetic plates
Magnetic plates

Super magnetic plate, 8×12 cm

Price
SFr. 363.68
Log in per disponibilità
1 * 1 ST
Product Details & Documents

Optimised magnetic plates with tailored properties especially designed for Magnetofection™.

  • Can be easily cleaned and decontaminated with 70% ethanol
  • Can be used at room temperature, 37 °C, +4 °C, etc.
  • Compatible with culture plates from most common suppliers
  • Solid, completely reusable

Suitable for all Magnetofection™ reagents, all cell culture dishes and flasks and can be used within incubators and with robots.

Specifiche
  • Descrizione:
    Super magnetic plate, 8×12 cm
  • Seq:
    01