Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Recycling Bins

Recycling bin, blu, 80 L, 380×500×800 mm
Codice catalogo LAMA1289BL
Nome del fornitore:   LAMAPLAST
Recycling Bins
Codice catalogo LAMA1289BL
Nome del fornitore:  LAMAPLAST
Recycling Bins
Recycling Bins

Recycling bin, blu, 80 L, 380×500×800 mm

Price
SFr. 268.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents

Square garbage bin with sack holder.

  • Made by HDPE
  • Complimentary pedal and wheel kit

Delivery information: Supplied with stickers for waste sorting: PLASTIC - GLASS - PAPER - ORGANIC WASTE.

Specifiche
  • Capacità:
    80 L
  • Colore:
    blu
  • Descrizione:
    Recycling bin
  • Seq:
    01
  • L×P×A:
    380×500×800 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.