Wringers for mops
Stainless steel flat mop wringer for VertiKlean® MAX™ and QuickTask™ mops, for 20 l buckets, 546×146×64 mm
Codice catalogo CONT2691
Nome del fornitore: CONTEC
Vai a
Product Details & Documents
Variety of wringers for curved and flat mops.
- Made of stainless steel
- Can be used with curved, flat, and string mops
- Ideal for use with Contec’s 15, 20, and 25 L bucket systems
- Fully autoclavable
Specifiche
- Descrizione:Stainless steel flat mop wringer for VertiKlean® MAX™ and QuickTask™ mops, for 20 l buckets
- Dimensioni:546×146×64 mm
- Seq:04