Safety spectacles, Solus™ 1000
Anti-UV (2C-1.2), anti-scratch, anti-fog, verde/nero, Amber PC
Codice catalogo 111-2378
Nome del fornitore: 3M
Vai a
Product Details & Documents
3M™ Solus™ 1000 series safety glasses feature a strong polycarbonate lens in a slim, stylish frame. These glasses feature a Scotchgard™ Anti-fog/anti-scratch coating, and a soft temple and nose bridge.
- Stylish and modern slim frame with colour accented soft temple components
- Strong polycarbonate lens for impact resistance
- Scotchgard™ Anti-Fog coating offers superior anti-fog and anti-scratch properties, even after washing multiple times
- Lightweight with soft cushioning on nose bridge
- Suitable for automatic cleaning, disinfection, and sterilisation by autoclaving at 121 °C
- Inner padding on temples for increased comfort and stability
- Excellent protection against ultraviolet (UV) radiation
Optional: Adjustable headband and foam or TPE gasket.
Certifications: EN 166
Documents
Specifiche
- Tipo:Anti-UV (2C-1.2), anti-scratch, anti-fog
- Colore:verde/nero
- Materiale lente:Amber PC
Accessories
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Foam insert for 3M™ Solus™ 1000 spectacles, for a closer fit to the faceFoam insert for 3M™ Solus™ 1000 spectacles, for a closer fit to the faceMMMASOLUSSRLog in per disponibilità1 * 5 ST - SFr. 40.70