Orders LinkContactUsLinkComponent
Boc-D-Pro-OH 97%
Bottle, 100 g
  APOSOR307074-100G
Boc-D-Pro-OH 97%
  APOSOR307074-100G
 :  37784-17-1
Boc-D-Pro-OH 97%
Bottle, 100 g
 :  37784-17-1
SFr. 34.70
1 * 100 g
 :  (2R)-1-[(tert-butoxy)carbonyl]pyrrolidine-2-carboxylic acid, N-Boc-D-proline
  •  :37784-17-1
  •  :C₁₀H₁₇NO₄
  •  :MFCD00063226
  •  :215.25 g/mol
  •  :134…137 °C
  •  :Refrigerata
  • Tipo di conf.:
    Bottle
  • Conf:
    100 g