Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Weighing boats, rounded

Weighing boats, rounded

0,5 ml, 50 mm
Codice catalogo NEUB1261-17203
Nome del fornitore:   NEUBERT VOLUME GLASSWAERE
Weighing boats, rounded
Codice catalogo NEUB1261-17203
Nome del fornitore:  NEUBERT VOLUME GLASSWAERE
Weighing boats, rounded

0,5 ml, 50 mm

Price
SFr. 60.80
Log in per disponibilità
1 * 1 pezzi
Product Details & Documents

Weighing funnels, borosilicate glass 3.3, designed to weigh chemicals or for weighing chemicals with tubular stalk.

  • Excellent chemical resistance
  • Excellent temperature Resistance
Specifiche
  • Capacità:
    0,5 ml
  • Lunghezza:
    50 mm
  • Seq:
    01