Fluorite Objectives for Revolve Microscope
Fluorite phase objective, 4×, 17 mm
Catalog # 630-3052
Supplier: DISCOVER ECHO
Jump to:
Product Details & Documents
Documents
ECHO Meet Revolve
Product Literature
Specifications
- Ingrandimento:4×
- Descrizione:Fluorite phase objective
- Seq:29
- Distanza di lavoro:17 mm