Fluorite Objectives for Revolve Microscope
Fluorite ELWD objective, 40×, 2,7 - 4,0 mm
N° de catalogue 630-3059
Fournisseur: DISCOVER ECHO
Aller à
Product Details & Documents
Documents
ECHO Meet Revolve
Product Literature
Spécifications
- Ingrandimento:40×
- Descrizione:Fluorite ELWD objective
- Seq:19
- Distanza di lavoro:2,7 - 4,0 mm