Fluorite Objectives for Revolve Microscope
Flourite ELWD phase objective, 20×, 6,6 - 7,8 mm
Catalog # 630-3056
Supplier: DISCOVER ECHO
Jump to:
Product Details & Documents
Specifications
- Ingrandimento:20×
- Descrizione:Flourite ELWD phase objective
- Seq:24
- Distanza di lavoro:6,6 - 7,8 mm