Orders LinkContactUsLinkComponent
Fluorite Objectives for Revolve Microscope
Fluorite oil phase objective, 60×, 0,12 mm
Katalognr. 630-3066
Anbieter:   DISCOVER ECHO
Fluorite Objectives for Revolve Microscope
Katalognr. 630-3066
Anbieter:  DISCOVER ECHO
Fluorite Objectives for Revolve Microscope
Fluorite oil phase objective, 60×, 0,12 mm
Preis
4 350.00 SFr.
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Technische Daten
  • Ingrandimento:
    60×
  • Descrizione:
    Fluorite oil phase objective
  • Seq:
    02
  • Distanza di lavoro:
    0,12 mm