FR Syringe pump holds 1 syringe from micro-liter sizes up to 60 ml (140 ml partially filled) infusion rates from 0.73 µl/hr (1 ml syringe) to 2100 ml/hr (60 ml syring
Codice catalogo MSPPNE-1000
Nome del fornitore: MARKET SOURCE PART PROCESS
Frequently Bought Together