Colour coded inserts, CapInsert™
rosso
Catalog # SIMPM957R
Supplier: Simport Scientific
Jump to:
Product Details & Documents
These colour coded inserts could be applied to the tube closure, which eliminates jeopardising the integrity of your sample by transferring it to another vial.
- Manually press fit and locks into place on top of closure
Specifications
- Colore:rosso
- Seq:04