Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Colesterolo NF

Colesterolo NF

Glass, amber, 25 g
Codice catalogo SPCMCH135-25GM
Nome del fornitore:   Spectrum Chemical
Colesterolo NF
Codice catalogo SPCMCH135-25GM
Nome del fornitore:  Spectrum Chemical
Numero CAS:  57-88-5
Colesterolo NF

Glass, amber, 25 g

Numero CAS:  57-88-5
Price
SFr. 119.00
Log in per disponibilità
1 * 25 g
Product Details & Documents

BSE/TSE statement (available upon request) Allergen statement (available upon request)

All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 
BSE/TSE statement (available upon request)
Allergen statement (available upon request)

Caution: LIGHT SENSITIVE: Keep tightly closed in light-resistant containers.

Sinonimi:  Cholest-5-en-3-ol, Cholesterin, (3Beta)-Cholest-5-en-3-ol
  • Numero CAS:57-88-5
  • Formula:C₂₇H₄₆O
  • Numero MDL:MFCD00003646
  • Molecular Weight:386.66 g/mol
  • IndexMerck:13,02221
  • Punto di ebollizione:360 °C (1013 hPa)
  • Punto di fusione:147…150 °C
  • Densità:1,07 g/cm³ (20 °C)
  • temperatura di conservazione:Ambiente
  • EINECS:200-353-2
Specifiche

Risultati dei test sulle specifiche

  • ASSAY (DRIED BASIS):
    95.0 - 102.0 %
  • MELTING RANGE:
    147° - 150°C
  • SPECIFIC ROTATION:
    -34° to -38°
  • SOLUBILITY IN ALCOHOL:
    No deposit or turbidity forms
  • ACIDITY:
    max 0.3 ml
  • LOSS ON DRYING:
    max 0.3%
  • RESIDUE ON IGNITION:
    max 0.1%
  • β-CHOLESTANOL (5a-CHOLESTAN-3β-ol,DIHYDROCHOLESTEROL):
    max 0.6 %
  • DESMOSTEROL:
    max 4%
  • LATHOSTEROL:
    max 2%
  • 24-DEHYDROLATHOSTEROL (5a-CHOLESTA-7,24 DIEN--3β-ol ):
    max 0.2 %
  • 4-METHYLCHOLEST-5-EN-3β-OL:
    max 0.5 %
  • TOTAL IMPURITIES INCLUDING RELATED STEROLS:
    max 5%
  • ELEMENTAL IMPURITIES:
    AS REPORTED
  • IDENTIFICATION (A):
    SPECTRUM MATCHES REFERENCE
  • IDENTIFICATION (B):
    -34° to -38°
  • IDENTIFICATION (C):
    147 - 150 °C

Specifiche

  • Tipo di conf.:
    Glass, amber
  • Conf:
    25 g

Specifiche