Electronic crimping tools
Electronic crimper for 11 mm aluminium crimp caps
Codice catalogo MANA735511
Nome del fornitore: MACHEREY-NAGEL
Vai a
Product Details & Documents
These battery-powered electronic crimping tools are suitable for aluminium crimp and Flip Top / Flip Off caps (not suitable for 20 mm magnetic / bi-metal crimp caps).
- Mobile tools for consistent and reproducible crimping results
- Crimping pressure adjustable by the control unit on top of the tool
- Long lasting lithium ion cell batteries (full battery charge for several hundred vials, life time of battery >1500 charges)
Certifications: CE certificate of conformity along with one year warranty.
Documents
Brochure-Crimping-tools-EN.pdf Brochure-Vials-and-caps-EN.pdf
Ulteriori dettagli sul prodotto
Specifiche
- Descrizione:Electronic crimper for 11 mm aluminium crimp caps
- Seq:01
Accessories
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Rack for 2 electronic crimping tools
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Replacement battery 6,6 V, 8,6 Wh