Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
QUANTOFIX® Test Strips, Swimming Pool Test

QUANTOFIX® Test Strips, Swimming Pool Test

Swimming pool test 5 in 1, 0 • 0.5 • 1 • 3 • 5 • 10 - mg/L - Cl₂ ; 0 • 80 • 120 • 180 • 240 - mg/L - CaCO₃ ; 6.4 • 6.8 • 7.5 • 7.6 • 8.4 ; 0 • 1 • 3 • 5 • 10 - mg/L - Cl₂, Alcalinity: Reaction with an acid and mixture of pH indicators; Free chlorine/ total chlorine: Colour reaction with azines; pH: Colour reaction of a pH indicator; Total hardness: Colour reaction with complex indicator, 40×40×120 mm (package), 50 test(s), 1
Codice catalogo MANA90759
Nome del fornitore:   MACHEREY-NAGEL
QUANTOFIX® Test Strips, Swimming Pool Test
Codice catalogo MANA90759
Nome del fornitore:  MACHEREY-NAGEL
Numero CAS:  
QUANTOFIX® Test Strips, Swimming Pool Test
QUANTOFIX® Test Strips, Swimming Pool Test

Swimming pool test 5 in 1, 0 • 0.5 • 1 • 3 • 5 • 10 - mg/L - Cl₂ ; 0 • 80 • 120 • 180 • 240 - mg/L - CaCO₃ ; 6.4 • 6.8 • 7.5 • 7.6 • 8.4 ; 0 • 1 • 3 • 5 • 10 - mg/L - Cl₂, Alcalinity: Reaction with an acid and mixture of pH indicators; Free chlorine/ total chlorine: Colour reaction with azines; pH: Colour reaction of a pH indicator; Total hardness: Colour reaction with complex indicator, 40×40×120 mm (package), 50 test(s), 1

Numero CAS:  
Price
36.40 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents

Semi-qualitative test strips for water qualities in swimming pool.

  • Easy dip-and-read procedure Multi-stage colour scale Provides reliable results within 30 seconds No accessories necessary for evaluation Desiccant in the stopper of tube container for optimal protection of the strips against humidity

These strips are used for quick and easy determination of water qualities in swimming pool. The test provides valuable results about the water hardness, the total alkalinity, free and total chlorine and pH.

Specifiche

Specifiche

  • Metodo chimico:
    Alcalinity: Reaction with an acid and mixture of pH indicators; Free chlorine/ total chlorine: Colour reaction with azines; pH: Colour reaction of a pH indicator; Total hardness: Colour reaction with complex indicator
  • Conf:
    1
  • Descrizione:
    Swimming pool test 5 in 1
  • Dimensioni:
    40×40×120 mm (package)
  • Test per kit:
    50 test(s)
  • Range di misura:
    0 • 0.5 • 1 • 3 • 5 • 10 - mg/L - Cl₂ ; 0 • 80 • 120 • 180 • 240 - mg/L - CaCO₃ ; 6.4 • 6.8 • 7.5 • 7.6 • 8.4 ; 0 • 1 • 3 • 5 • 10 - mg/L - Cl₂
  • Cat. No.:
    MANA90759
  • Contenuti:
    50 test strips in a tube
  • Peso:
    35,5 g
  • Strumento:
    No
  • Parametro:
    All reagents included
  • Conformità:
    Not CE certified and no hazardous material
  • Piattaforma:
    Semi-quantitative test strips
  • Durata a magazzino (mesi):
    24 (from production)

Specifiche

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.