ContactUsLinkComponent
Recombinat Protein,Human Frizzled-2 FZD2 Protein, Fc Tag,Human Frizzled-2, Fc Tag (FZ2-H5251) is expressed from human 293 cells (HEK 293). It contains AA Gln 24 - Ser 156 (Accession AAI13403).This protein carries a human IgG1 Fc tag at the C-terminus 1 * 100 µG
Catalog # ACRBFZ2-H5251100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human Frizzled-2 FZD2 Protein, Fc Tag,Human Frizzled-2, Fc Tag (FZ2-H5251) is expressed from human 293 cells (HEK 293). It contains AA Gln 24 - Ser 156 (Accession AAI13403).This protein carries a human IgG1 Fc tag at the C-terminus 1 * 100 µG
Catalog # ACRBFZ2-H5251100UG
Supplier:  ACRO Biosystems
Frequently Bought Together