Inspection date labels
Inspection date labels, Vinile, 2028, verde, 15 mm
Codice catalogo BRDY834118
Nome del fornitore: Brady
Vai a
Product Details & Documents
These inspection date labels are made of vinyl cloth. They can be used wherever a frequent and regular inspection of machinery and equipment is required. Simply mark the label with a hole punch and apply wherever you need it.
- Show evidence of unauthorised removal
Use the yearly colour system to easily identify when your equipment was last inspected.
Documents
Specifiche
- Colore:verde
- Descrizione:Inspection date labels
- Dimensioni:15 mm
- Materiale:Vinile
- Seq:13
- Legend:2028