Kits for Precellys®
Kit Precellys® Ceramic da 2,8 mm, rinforzato, 50 provette da 2 ml, preriempite con sferette in ceramica, Tessuti duri, CK28-R/ 2 ml
Nome del fornitore: Bertin Instruments
Vai a
Product Details & Documents
The Precellys lysing kits offer the best versatility of volumes and grinding material to process any type of samples: Human, animal, plants, microorganisms or soil.
- More than 30 different lysing matrixes
- Reinforced tubes are suitable for use at the maximum energy levels
- Easy-handling features allow the Precellys to be used without any advanced training
These kits are specifically for sample preparation for Precellys homogenisers. They can process any type of sample. Choosing the appropriate lysing kit maximises the quantity and quality of extraction by decreasing the process time and limiting the heat generated.
Reinforced tubes have thicker walls and their top contains an o-ring to make it more resistant.
Documents
Certificates
Cerca per numero di lottoBertin-Precellys-Lysing-Kits Katalogseite Precellys Aufschluss-Kits
Letteratura sul prodotto
BERTP000913-LYSK0A_240627-890.pdf BERTP000947-LYSK0A_240716-830.pdf CoC_BERTP000913-LYSK0A_200707-804 CoC_BERTP000913-LYSK0A_210809-890 coc_BERTP000916-LYSK0A_250716-830
More
Hair Grinding In 7 ml Tube Hard Sample Grinding with 7 ml Metal Tube Improving Compound Extraction Efficiency using the Precellys24 Protein Extraction From E.coli Using Precellys® Evolution Quantative recovery of BMP-2 after grinding of demineralised bone with Precellys® Evolution
More
SDS_IT_CH_BERTP000911-LYSK0A SDS_IT_CH_BERTP000912
Scheda di sicurezza
Usage limits of Precellys lysing kits
Scheda dati
Specifiche
- Descrizione:Kit Precellys® Ceramic da 2,8 mm, rinforzato, 50 provette da 2 ml, preriempite con sferette in ceramica
- Seq:022
- Tipo:CK28-R/ 2 ml
- Per:Tessuti duri