Orders LinkContactUsLinkComponent
High content cell imaging system, ImageXpress Micro 4
Filter set, Cy3
  MLDV5039858
High content cell imaging system, ImageXpress Micro 4
  MLDV5039858
High content cell imaging system, ImageXpress Micro 4
High content cell imaging system, ImageXpress Micro 4
Filter set, Cy3
SFr. 2’629.00
1 * 1 ST
The ImageXpress Micro 4 high content imaging system represents the culmination of four generations of imaging expertise. The agile design allows you to boost your research with this faster than ever system, while providing the option to upgrade to confocal in the future to align with your research needs.
  • Tackle a wide range of assays with oil and air objectives ranging from 1× to 100×
  • Automated five-position filter cube changer to facilitate multiplex assays with up to five filter sets and four objectives for a single experiment
  • Optically-encoded linear voice coil driven stages with encoders that measure position with better than 25 nm resolution, to achieve high accuracy and precision in X, Y and Z axes
  • Expand your research capabilities with environmental control: Temperature, humidity and CO₂ control for multi-day, live cell experiments), transmitted light true phase contrast, confocal imaging (field upgradable), and fluidics (onboard disposable pipetting)
Capture images of whole organisms and cellular or intracellular events with the ImageXpress Micro 4 system, configured to suit your specific biological needs. We offer unlimited configurability with user exchangeable filter cubes, a wide range of objective lenses and environmental control, transmitted light, confocal imaging and fluidic options. Speed up your research by acquiring more than 245000 wells per day at 4× magnification with the system’s unique multi-well crop feature, and resolve the fine detail you need with increasing magnifications.
Combined with MetaXpress high content image acquisition and analysis software, the ImageXpress Micro 4 system is an end-to-end solution to interpret images, understand data and explore ideas.
  • Descrizione:
    Filter set, Cy3
  • Seq:
    106
Complementary Products
  •  
    Conf
    Descrizione
    Sterilità
    Formato del siero
    Colore
    Volume di lavoro raccomandato
    Sterili
    Confezione
    Superficie coltura
    Volume pozzetti
  • 30026.
    1 KIT
    Calcein-AM Cell Viability Assay kit, 1000 assays
    1 * 1 KIT - 501.00 SFr.
  • 30029.
    1 KIT
    Kit per analisi di caspasi 3 NucView™ 488 per cellule vive, 100 test
    1 * 1 KIT - 758.00 SFr.
  • S1810-500
    Sterile filtered
    Foetal bovine serum, South American origin
    15
    1 * 500 mL - 330.00 SFr.
  • 100.103.010
    1
    GrowDex®, 10 ml syringe
    1 * 1 ST - 348.00 SFr.
  • 30002.
    1 KIT
    Kit di campionatura della viabilità/citotossicità per cellule animali vive e morte, 300 campionature
    1 * 1 KIT - 528.00 SFr.
  • 30009-2
    1 KIT
    Caspase-3 DEVD-R110 Fluorometric HTS Assay Kit, 10 ml
    1 * 1 KIT - 354.00 SFr.
  • 734-2327
    trasparente
    0,075 - 0,20 ml
    1/Sacchetto
    0,33 cm²
    96-piastre a pozzetti, fondo piatto, trattate con TC
    17
    1 * 100 ST - 305.00 SFr.
  • 735-0523
    Microlite™ 1+ strisce, 1×12/strip
    1 * 320 ST - 252.00 SFr.
  • CORN3904BC
    nero
    +
    360 µl
    96-well microplate, clear flat bottom, tissue culture treated, with lid
    75 - 200 µl
    21
    1 * 100 ST - 1 380.00 SFr.
  • 734-2097
    trasparente
    200 µl
    06
    1 * 50 ST - 271.00 SFr.