Orders LinkContactUsLinkComponent
Probes for testo 440
Temperature and humidity probe head, 160×28×28 mm, diameter probe head: 12 mm, length probe shaft: 140 mm, −20...+70 °C
RH: 0 - 100 %, T: 0,5 °C
RH: ±2% (5 - 90%); ±0,03% RH/K (k=1), T: 0,1 °C
RH: 0,1 %
Codice catalogo TEST0636.9730
Nome del fornitore:   Testo
Probes for testo 440
Codice catalogo TEST0636.9730
Nome del fornitore:  Testo
Probes for testo 440
Probes for testo 440
Temperature and humidity probe head, 160×28×28 mm, diameter probe head: 12 mm, length probe shaft: 140 mm, −20...+70 °C
RH: 0 - 100 %, T: 0,5 °C
RH: ±2% (5 - 90%); ±0,03% RH/K (k=1), T: 0,1 °C
RH: 0,1 %
Price
126.00 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Use the humidity/temperature probes (with an appropriate measuring instrument) for the reliable measurement of relative humidity and temperature. Humidity sensor is durable and traceable to international humidity standards, such as ILAC, PTB and NIST. Thus, even after years of use you can still count on measurement results that are reliable and correct.
Specifiche
  • Range di misura:
    −20...+70 °C
    RH: 0 - 100 %
  • Risoluzione:
    T: 0,1 °C
    RH: 0,1 %
  • Descrizione:
    Temperature and humidity probe head, 160×28×28 mm, diameter probe head: 12 mm, length probe shaft: 140 mm
  • Seq:
    23
  • Accuratezza:
    T: 0,5 °C
    RH: ±2% (5 - 90%); ±0,03% RH/K (k=1)