Labels, hazard warning
Hazard labels: general warning, PIC W001-TRI 025-PE-ROLL/1, Poliestere, nero on giallo, 25×18 mm, 250 labels/roll
Codice catalogo BRDY826610
Nome del fornitore: Brady
Vai a
Product Details & Documents
These self adhesive signs are designed to warn people to be careful and to take precautions of potential hazards.
Available in different sizes.
Certifications: Conforms to Health and Safety (Safety Signs and Signals) Regulations 1996.
Specifiche
- Colore:nero on giallo
- Descrizione:Hazard labels: general warning
- Dimensioni:25×18 mm
- Materiale:Poliestere
- Seq:03
- Modello:PIC W001-TRI 025-PE-ROLL/1
- Confezione:250 labels/roll