Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Human recombinant SMAC

Human recombinant SMAC

5 µG
Codice catalogo APOSBITP2127-5UG
Nome del fornitore:   Apollo Scientific
Human recombinant SMAC
Codice catalogo APOSBITP2127-5UG
Nome del fornitore:  Apollo Scientific
Numero CAS:  
Human recombinant SMAC

5 µG

Numero CAS:  
Price
SFr. 191.00
Log in per disponibilità
1 * 5 µG
Product Details & Documents
Specifiche

Specifiche

  • Conf:
    5 µG
  • Cat. No.:
    APOSBITP2127-5UG
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    SMAC
  • Protein synonyms:
    DIABLO
  • Specie:
    Human

Specifiche

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.