Orders LinkContactUsLinkComponent
Heller's basal salt mixture
Heller's basal salt mixture
Heller's basal salt mixture 10×1 L, 10
Catalog # APOSPMH393-10X1L
Supplier:   Apollo Scientific
Heller's basal salt mixture
Catalog # APOSPMH393-10X1L
Supplier:  Apollo Scientific
CAS Number:  
Heller's basal salt mixture
Heller's basal salt mixture 10×1 L, 10
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 10 ST
Product Details & Documents
Contains macro/micronutrients as described by Heller, 1953 Plant tissue culture testedFull specification given in appendix.
  • MFCD00283413
Specifications
Specifications
  • Conf:
    10
  • Descrizione:
    Heller's basal salt mixture 10×1 L