Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

D-(-)-Penicillamine

Bottle, 25 g
Catalog # APOSOR18365-25G
Irritant
Supplier:   Apollo Scientific
D-(-)-Penicillamine
Catalog # APOSOR18365-25G
Supplier:  Apollo Scientific
CAS Number:  52-67-5
D-(-)-Penicillamine

Bottle, 25 g

CAS Number:  52-67-5
Price
SFr. 108.00
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  H-D-Pen-OH, (S)-2-Amino-3-mercapto-3-methylbutanoic acid, Penicillamine
  • CAS Number:52-67-5
  • Formula:C₅H₁₁NO₂S
  • MDL Number:MFCD00064302
  • Molecular Weight:149.2 g/mol
  • Melting Pt:∼ 212 °C (dec.)
  • Melting Pt:ca 212 °C (dec.)
  • Storage Temperature:Refrigerata
Specifications

Specifications

  • Tipo di conf.:
    Bottle
  • Conf:
    25 g

Specifications